Assay Details
Target Gene Details
Entrez Gene ID: | 26876 |
Gene Name: | alcohol dehydrogenase 4 (class II), pi polypeptide |
Gene Aliases: |
Adh2 |
Location: |
Chr.3:138415497-138430892 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adh4 | NM_011996.2 | NP_036126.2 | ||
AJ245750.1 | CAB57455.1 | |||
AK132994.1 | BAE21459.1 | |||
BC100729.1 | AAI00730.1 | |||
BC100730.1 | AAI00731.1 | |||
BC100736.1 | AAI00737.1 |
More Information
Set Membership: |
Intragenic Non-exonic |