Assay Details
Target Gene Details
Entrez Gene ID: | 56046 |
Gene Name: | ubiquinol-cytochrome c reductase complex assembly factor 1 |
Gene Aliases: |
2310079L17Rik, 2410003P15Rik, 3110038N19Rik, Bfzb, Bfzp, Cbp3, Uqcc |
Location: |
Chr.2:155846886-155930343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uqcc1 | NM_018888.4 | NP_061376.2 | ||
NR_024487.2 | ||||
AF253516.1 | AAF64519.1 | |||
AK010226.1 | 1 | 110 | ||
AK010376.1 | BAB26894.1 | |||
AK011152.1 | BAB27433.1 | |||
AK011953.1 | ||||
AK019425.1 | ||||
AK054294.1 | 3 | 264 | ||
AK077323.1 | BAC36749.1 | |||
AK139488.1 | BAE24034.1 | |||
AK169413.1 | BAE41159.1 | |||
BC050884.1 | AAH50884.1 | |||
BC057570.1 | AAH57570.1 |
More Information
Set Membership: |
Intragenic Non-exonic |