Assay Details
Target Gene Details
Entrez Gene ID: | 11722 |
Gene Name: | amylase 1, salivary |
Gene Aliases: |
Amy-1, Amy1a, C030014B17Rik |
Location: |
Chr.3:113555952-113577750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Amy1 | NM_001110505.1 | NP_001103975.1 | ||
NM_007446.2 | NP_031472.2 | |||
AA616210.1 | ||||
AK029642.1 | BAC26543.1 | |||
BC004698.1 | AAH04698.1 | |||
BC009121.1 | AAH09121.1 | |||
J00356.1 | AAA37221.1 | |||
V00717.1 | CAA24097.1 | |||
V00719.1 | CAA24099.1 | |||
V00720.1 | CAA24100.1 |
More Information
Set Membership: |
Intragenic Non-exonic |