Assay Details
Target Gene Details
Entrez Gene ID: | 57432 |
Gene Name: | zinc finger CCCH type containing 8 |
Gene Aliases: |
AU020882, E130108N08Rik, Fliz1, Zc3hdc8 |
Location: |
Chr.2:128926268-128944084 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zc3h8 | NM_020594.2 | NP_065619.2 | ||
AF061961.1 | AAF74513.1 | |||
AK132679.1 | BAE21298.1 | |||
AK164491.1 | BAE37808.1 | |||
BC048687.1 | AAH48687.1 |
More Information
Set Membership: |
Intragenic Non-exonic |