Assay Details
Target Gene Details
Entrez Gene ID: | 320024 |
Gene Name: | neutral cholesterol ester hydrolase 1 |
Gene Aliases: |
Aadacl1, B230106I24Rik, CPO-BP, Nceh |
Location: |
Chr.3:27183004-27245266 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nceh1 | NM_178772.3 | NP_848887.1 | ||
AK034339.1 | BAC28680.1 | |||
AK045363.1 | BAC32327.1 | |||
AK085767.1 | ||||
AK128968.1 | ||||
AK135837.1 | BAE22685.1 | |||
AK173158.1 | BAD32436.1 | |||
BC082569.1 | AAH82569.1 |
More Information
Set Membership: |
Intragenic Non-exonic |