Assay Details
Target Gene Details
Entrez Gene ID: | 17183 |
Gene Name: | matrilin 4 |
Gene Aliases: |
matrilin-4 |
Location: |
Chr.2:164389393-164405160 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Matn4 | NM_001252563.1 | NP_001239492.1 | ||
NM_013592.4 | NP_038620.2 | |||
AJ006140.1 | CAA06889.1 | |||
AK041073.1 | ||||
AK160786.1 | ||||
BB664034.1 | ||||
BC036558.1 | AAH36558.1 |
Target Gene Details
Entrez Gene ID: | 19668 |
Gene Name: | recombination signal binding protein for immunoglobulin kappa J region-like |
Gene Aliases: |
RBP-L, Rbpsuhl |
Location: |
Chr.2:164403056-164415448 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbpjl | NM_009036.1 | NP_033062.1 | ||
AK052449.1 | ||||
AK131814.1 | BAE20815.1 | |||
AK131832.1 | ||||
BC152311.1 | ||||
Y10926.1 | CAA71856.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |