Assay Details
Target Gene Details
Entrez Gene ID: | 27054 |
Gene Name: | SEC23B (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.2:144556229-144590753 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec23b | NM_001252543.1 | 5 | 594 | NP_001239472.1 |
NM_001252544.1 | 5 | 587 | NP_001239473.1 | |
NM_001252545.1 | 5 | 576 | NP_001239474.1 | |
NM_001281816.1 | 2 | 216 | NP_001268745.1 | |
NM_019787.4 | 5 | 605 | NP_062761.2 | |
AF200326.1 | 4 | 477 | AAF08301.1 | |
AK014597.1 | 5 | 555 | BAB29452.1 | |
AK029661.1 | 2 | 217 | BAC26553.1 | |
AK139729.1 | 5 | 519 | ||
AK146083.1 | 5 | 554 | BAE26887.1 | |
AK149693.1 | 5 | 534 | BAE29030.1 | |
AK153214.1 | 5 | 561 | BAE31812.1 | |
AK155168.1 | 5 | 535 | BAE33090.1 | |
AK158398.1 | 5 | 572 | BAE34488.1 | |
AK166330.1 | 5 | 537 | BAE38710.1 | |
AK167382.1 | 5 | 543 | ||
AK167732.1 | 5 | 523 | BAE39773.1 | |
AK171597.1 | 5 | 535 | BAE42552.1 | |
AK207187.1 | 1 | 85 | ||
BC005464.1 | 5 | 530 | AAH05464.1 | |
BC011160.1 | 5 | 557 | AAH11160.1 |
More Information
Set Membership: |
Intragenic Exonic |