Assay Details
Target Gene Details
Entrez Gene ID: | 56878 |
Gene Name: | RNA binding motif, single stranded interacting protein 1 |
Gene Aliases: |
2600014B10Rik, AI255215, MSSP-1, MSSP-2, MSSP-3, YC1 |
Location: |
Chr.2:60750193-60963450 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rbms1 | NM_001141931.1 | NP_001135403.1 | ||
NM_001141932.1 | NP_001135404.1 | |||
NM_020296.2 | NP_064692.2 | |||
AB026569.1 | BAA77262.1 | |||
AK133054.1 | BAE21490.1 | |||
AK135280.1 | BAE22471.1 | |||
AK145897.1 | BAE26733.1 | |||
AK147109.1 | BAE27681.1 | |||
AK161102.1 | BAE36195.1 | |||
AK161416.1 | BAE36384.1 | |||
AK164307.1 | BAE37728.1 | |||
BC016501.1 | ||||
BC056609.1 | AAH56609.1 | |||
BC057866.1 | AAH57866.1 |