Assay Details
Target Gene Details
Entrez Gene ID: | 27054 |
Gene Name: | SEC23B (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.2:144556229-144590753 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec23b | NM_001252543.1 | NP_001239472.1 | ||
NM_001252544.1 | NP_001239473.1 | |||
NM_001252545.1 | NP_001239474.1 | |||
NM_001281816.1 | NP_001268745.1 | |||
NM_019787.4 | NP_062761.2 | |||
AF200326.1 | AAF08301.1 | |||
AK014597.1 | BAB29452.1 | |||
AK029661.1 | BAC26553.1 | |||
AK139729.1 | ||||
AK146083.1 | BAE26887.1 | |||
AK149693.1 | BAE29030.1 | |||
AK153214.1 | BAE31812.1 | |||
AK155168.1 | BAE33090.1 | |||
AK158398.1 | BAE34488.1 | |||
AK166330.1 | BAE38710.1 | |||
AK167382.1 | ||||
AK167732.1 | BAE39773.1 | |||
AK171597.1 | BAE42552.1 | |||
AK209568.1 | ||||
AK220080.1 | ||||
BC005464.1 | AAH05464.1 | |||
BC011160.1 | AAH11160.1 | |||
BU614902.1 | ||||
CF724836.1 |
More Information
Set Membership: |
Intragenic Non-exonic |