Assay Details
Target Gene Details
Entrez Gene ID: | 20811 |
Gene Name: | src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites |
Gene Aliases: |
A230069J08Rik, srm |
Location: |
Chr.2:181205562-181218783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srms | NM_011481.3 | NP_035611.3 | ||
AK049582.1 | BAC33825.1 | |||
BC120633.1 | ||||
BC125325.1 | ||||
D26186.1 | BAA05331.1 | |||
D49427.1 | BAA08406.1 |