Assay Details
Target Gene Details
Entrez Gene ID: | 20568 |
Gene Name: | secretory leukocyte peptidase inhibitor |
Gene Aliases: |
- |
Location: |
Chr.2:164354070-164356507 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slpi | NM_011414.3 | NP_035544.1 | ||
AK010170.1 | ||||
BC028509.1 | AAH28509.1 | |||
U88093.1 | AAC53140.1 | |||
U94341.1 | AAC53394.1 |
More Information
Set Membership: |
Intragenic Non-exonic |