Assay Details
Target Gene Details
Entrez Gene ID: | 20014 |
Gene Name: | ribophorin II |
Gene Aliases: |
1300012C06Rik, AV261018, Rpn-2 |
Location: |
Chr.2:157279011-157326318 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpn2 | NM_019642.4 | 3 | 479 | NP_062616.2 |
AK004968.1 | 3 | 480 | BAB23707.1 | |
AK018761.2 | 3 | 518 | ||
AK153952.1 | 3 | 340 | BAE32275.1 | |
AK161624.1 | 3 | 290 | BAE36498.1 | |
AK166778.1 | 3 | 265 | BAE39013.1 | |
AK167864.1 | 3 | 261 | ||
AK167866.1 | 3 | 261 | ||
AK168221.1 | 3 | 261 | ||
AK212593.1 | ||||
BC010509.1 | 2 | 252 | AAH10509.1 | |
BC046806.1 | 3 | 507 | AAH46806.1 | |
D31717.1 | 2 | 265 | BAA06525.1 |
More Information
Set Membership: |
Intragenic Exonic |