Assay Details
Target Gene Details
Entrez Gene ID: | 20014 |
Gene Name: | ribophorin II |
Gene Aliases: |
1300012C06Rik, AV261018, Rpn-2 |
Location: |
Chr.2:157279011-157326318 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpn2 | NM_019642.4 | NP_062616.2 | ||
AK004968.1 | BAB23707.1 | |||
AK018761.2 | ||||
AK087923.1 | ||||
AK153952.1 | BAE32275.1 | |||
AK161624.1 | BAE36498.1 | |||
AK166778.1 | BAE39013.1 | |||
AK167864.1 | ||||
AK167866.1 | ||||
AK168221.1 | ||||
BC010509.1 | AAH10509.1 | |||
BC046806.1 | AAH46806.1 | |||
D31717.1 | BAA06525.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |