Assay Details
Target Gene Details
Entrez Gene ID: | 56086 |
Gene Name: | SET nuclear oncogene |
Gene Aliases: |
2610030F17Rik, 5730420M11Rik, AA407739, I-2PP2A, StF-IT-1, TAF-I |
Location: |
Chr.2:30061996-30072577 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Set | NM_001204875.1 | NP_001191804.1 | ||
NM_023871.4 | NP_076360.1 | |||
AB015613.1 | BAA34736.1 | |||
AB044937.1 | BAB20793.1 | |||
AK011630.1 | BAB27745.1 | |||
AK019960.1 | BAB31936.1 | |||
AK172327.1 | BAE42947.1 | |||
BC018255.1 | AAH18255.1 | |||
BC103791.1 | AAI03792.1 | |||
BQ899734.1 |
More Information
Set Membership: |
Intragenic Non-exonic |