Assay Details
Target Gene Details
Entrez Gene ID: | 227525 |
Gene Name: | DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae) |
Gene Aliases: |
9930121L06Rik, AI661365, Art, Snm1l |
Location: |
Chr.2:3424062-3474330 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dclre1c | NM_001110214.1 | 15 | 2417 | NP_001103684.1 |
NM_175683.4 | 16 | 2768 | NP_783614.1 | |
AK052369.1 | 16 | 2724 | BAC34960.1 | |
AK088810.1 | 14 | 2550 | BAC40586.1 |
Target Gene Details
Entrez Gene ID: | 64707 |
Gene Name: | suppressor of variegation 3-9 homolog 2 (Drosophila) |
Gene Aliases: |
4930507K23Rik, AA536750, D030054H19Rik, D2Ertd544e, KMT1B |
Location: |
Chr.2:3455815-3474986 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Suv39h2 | NM_022724.4 | 5 | 1301 | NP_073561.2 |
NR_027509.1 | 4 | 1155 | ||
AF149205.1 | 5 | 1279 | AAG09134.1 | |
AK015728.1 | 3 | 347 | BAB29948.1 | |
AK045206.1 | 4 | 1139 | ||
AK083457.1 | 5 | 1279 | BAC38921.1 | |
AK165278.1 | 4 | 954 | BAE38116.1 | |
BC032960.1 | 4 | 624 | AAH32960.1 |
More Information
Set Membership: |
Intragenic Exonic |