Assay Details
Target Gene Details
Entrez Gene ID: | 68365 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
0610030G24Rik, 2810475J17Rik, A830021G03Rik, AI314285, AI649155, D030017L14Rik |
Location: |
Chr.2:35180205-35201431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rab14 | NM_026697.3 | NP_080973.1 | ||
AB232609.1 | BAF02871.1 | |||
AJ968955.1 | CAI92125.1 | |||
AJ968956.1 | CAI92126.1 | |||
AJ968957.1 | CAI92127.1 | |||
AJ968958.1 | CAI92128.1 | |||
AJ968959.1 | CAI92129.1 | |||
AJ968960.1 | CAI92130.1 | |||
AK002704.1 | BAB22298.1 | |||
AK013410.1 | ||||
AK133963.1 | BAE21957.1 | |||
AK135751.1 | ||||
AK160859.1 | BAE36051.1 | |||
AK165812.1 | BAE38391.1 | |||
AK168551.1 | BAE40426.1 | |||
AK168867.1 | BAE40686.1 | |||
AK169396.1 | BAE41143.1 | |||
BC009085.1 | AAH09085.1 | |||
BC025139.1 | AAH25139.1 | |||
BC056648.1 | AAH56648.1 |