Assay Details
Target Gene Details
Entrez Gene ID: | 57138 |
Gene Name: | solute carrier family 12, member 5 |
Gene Aliases: |
KCC2, mKIAA1176 |
Location: |
Chr.2:164960818-164999731 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc12a5 | NM_020333.2 | NP_065066.2 | ||
AF332063.1 | AAK56092.1 | |||
AF332064.1 | AAK56093.1 | |||
AK122460.1 | BAC65742.1 | |||
AK147262.1 | BAE27805.1 | |||
AK165769.1 | BAE38372.1 | |||
BC054808.1 | AAH54808.1 | |||
KJ535320.1 | ||||
KJ535321.1 | ||||
KJ535322.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |