Assay Details
Target Gene Details
Entrez Gene ID: | 20687 |
Gene Name: | trans-acting transcription factor 3 |
Gene Aliases: |
D130027J01Rik |
Location: |
Chr.2:72936430-72980446 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sp3 | NM_001018042.3 | 5 | 1930 | NP_001018052.1 |
NM_001098425.1 | 3 | 1839 | NP_001091895.1 | |
AF062567.1 | 3 | 1566 | AAC16322.1 | |
AK004607.1 | 2 | 1580 | BAC25090.1 | |
AK088238.1 | 3 | 1814 | ||
AK132702.1 | 3 | 1585 | BAE21310.1 | |
BC027797.1 | 2 | 1052 | ||
BC079874.1 | 5 | 1930 | AAH79874.1 |
More Information
Set Membership: |
Intragenic Exonic |