Assay Details
Target Gene Details
Entrez Gene ID: | 110078 |
Gene Name: | brain glycogen phosphorylase |
Gene Aliases: |
- |
Location: |
Chr.2:150786796-150831748 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pygb | NM_153781.1 | NP_722476.1 | ||
AK032769.1 | BAE20482.1 | |||
AK035673.1 | ||||
AK133794.1 | BAE21845.1 | |||
AK134667.1 | BAE22233.1 | |||
AK142716.1 | ||||
AK147763.1 | BAE28122.1 | |||
AK168381.1 | ||||
AK169052.1 | BAE40840.1 | |||
AK169631.1 | ||||
AK178330.1 | ||||
AK184046.1 | ||||
AK213706.1 | ||||
BC032209.1 | AAH32209.1 | |||
BC035283.1 | AAH35283.1 |
More Information
Set Membership: |
Intragenic Non-exonic |