Assay Details
Target Gene Details
Entrez Gene ID: | 329384 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae) |
Gene Aliases: |
2210013M04Rik, AW046744 |
Location: |
Chr.2:32775738-32777593 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptrh1 | NM_178595.3 | 2 | 148 | NP_848710.1 |
AK075833.1 | 2 | 149 | BAC35994.1 | |
BC098218.1 | 2 | 188 | AAH98218.1 |
More Information
Set Membership: |
Intragenic Exonic |