Assay Details
Target Gene Details
Entrez Gene ID: | 54338 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 2 |
Gene Aliases: |
AI844736, NBTL1, SVCT2, Slc23a1, YSPL2, YSPL3, mKIAA0238 |
Location: |
Chr.2:132052496-132145108 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc23a2 | NM_018824.2 | NP_061294.2 | ||
AB038145.1 | BAA90751.1 | |||
AK087175.1 | BAC39819.1 | |||
AK169991.1 | BAE41501.1 | |||
AY004874.1 | AAG02252.1 | |||
BC050823.1 | AAH50823.1 |
More Information
Set Membership: |
Intragenic Non-exonic |