Assay Details
Target Gene Details
Entrez Gene ID: | 27387 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
Chat, Nsp3, Shep1 |
Location: |
Chr.2:32721055-32755007 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh2d3c | NM_001252547.1 | NP_001239476.1 | ||
NM_013781.3 | NP_038809.1 | |||
NR_045535.1 | ||||
AB030442.1 | BAA90557.1 | |||
AB043953.1 | BAA96361.1 | |||
AF168364.1 | AAF13305.1 | |||
AK042709.1 | BAC31340.1 | |||
AK155165.1 | BAE33088.1 | |||
AK172072.1 | ||||
BC113203.1 | AAI13204.1 |