Assay Details
Target Gene Details
Entrez Gene ID: | 66860 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 |
Gene Aliases: |
1200003E16Rik, Tanc, mKIAA1728 |
Location: |
Chr.2:59611848-59846213 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tanc1 | NM_001290659.1 | 16 | 2950 | NP_001277588.1 |
NM_198294.3 | 17 | 3126 | NP_938036.2 | |
AB093299.1 | 6 | 1219 | BAC41481.1 | |
AK030022.1 | 14 | 2730 | BAC26741.1 | |
BC079914.1 | 16 | 2929 | ||
BC141367.1 | 16 | 2940 | ||
CX769146.1 |