Assay Details
Target Gene Details
Entrez Gene ID: | 66860 |
Gene Name: | tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1 |
Gene Aliases: |
1200003E16Rik, Tanc, mKIAA1728 |
Location: |
Chr.2:59611848-59846213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tanc1 | NM_001290659.1 | NP_001277588.1 | ||
NM_198294.3 | NP_938036.2 | |||
AB093299.1 | BAC41481.1 | |||
AK030022.1 | BAC26741.1 | |||
AK134490.1 | BAE22161.1 | |||
BC079914.1 | ||||
BC141367.1 |