Assay Details
Target Gene Details
Entrez Gene ID: | 192166 |
Gene Name: | sarcosine dehydrogenase |
Gene Aliases: |
- |
Location: |
Chr.2:27188393-27248332 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sardh | NM_138665.2 | NP_619606.1 | ||
AK140451.1 | ||||
AK159236.1 | BAE34920.1 | |||
AK159681.1 | BAE35284.1 | |||
AK159704.1 | BAE35303.1 | |||
AK163515.1 | BAE37376.1 | |||
AK163660.1 | BAE37445.1 | |||
AK180924.1 | ||||
AK192092.1 | ||||
BC003456.1 | AAH03456.1 |
More Information
Set Membership: |
Intragenic Non-exonic |