Assay Details
Target Gene Details
Entrez Gene ID: | 192166 |
Gene Name: | sarcosine dehydrogenase |
Gene Aliases: |
- |
Location: |
Chr.2:27188393-27248332 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sardh | NM_138665.2 | 4 | 1275 | NP_619606.1 |
AK140451.1 | 3 | 551 | ||
AK159236.1 | 4 | 1239 | BAE34920.1 | |
AK159681.1 | 4 | 1239 | BAE35284.1 | |
AK159704.1 | 4 | 1239 | BAE35303.1 | |
AK163515.1 | 4 | 804 | BAE37376.1 | |
AK163660.1 | 3 | 551 | BAE37445.1 | |
BC003456.1 | 4 | 784 | AAH03456.1 |
More Information
Set Membership: |
Intragenic Exonic |