Assay Details
Target Gene Details
Entrez Gene ID: | 192166 |
Gene Name: | sarcosine dehydrogenase |
Gene Aliases: |
- |
Location: |
Chr.2:27188393-27248332 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sardh | NM_138665.2 | 15 | 2534 | NP_619606.1 |
AK087163.1 | BAC39816.1 | |||
AK140451.1 | 14 | 1811 | ||
AK159236.1 | 15 | 2498 | BAE34920.1 | |
AK159681.1 | 15 | 2498 | BAE35284.1 | |
AK159704.1 | 15 | 2498 | BAE35303.1 | |
AK163515.1 | 15 | 2063 | BAE37376.1 | |
AK163660.1 | 14 | 1810 | BAE37445.1 | |
AK176928.1 | 3 | 315 | ||
AK178634.1 | 3 | 282 | ||
AK182037.1 | 4 | 336 | ||
AK183193.1 | 3 | 270 | ||
AK208205.1 | 4 | 337 | ||
BC003456.1 | 15 | 2043 | AAH03456.1 |
More Information
Set Membership: |
Intragenic Exonic |