Assay Details
Target Gene Details
Entrez Gene ID: | 241274 |
Gene Name: | patatin-like phospholipase domain containing 7 |
Gene Aliases: |
BC027342, E430013P11Rik, Nre |
Location: |
Chr.2:24975171-25054072 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnpla7 | NM_146251.4 | NP_666363.3 | ||
AK038505.1 | ||||
AK088362.1 | BAC40302.1 | |||
AK148380.1 | BAE28519.1 | |||
AK170417.1 | BAE41783.1 | |||
BC025621.1 | AAH25621.1 | |||
BC141365.1 | ||||
BC141366.1 | ||||
EU293208.1 | ||||
EU368675.1 | ||||
EU402948.2 |
More Information
Set Membership: |
Intragenic Non-exonic |