Assay Details
Target Gene Details
Entrez Gene ID: | 18779 |
Gene Name: | phospholipase A2 receptor 1 |
Gene Aliases: |
Pla2-r, Pla2g1br, Pla2r |
Location: |
Chr.2:60417543-60553350 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2r1 | NM_008867.2 | 8 | 1554 | NP_032893.1 |
AK076366.1 | 8 | 1535 | ||
AK143853.1 | 8 | 1533 | ||
BC048780.1 | 8 | 1524 | AAH48780.1 | |
BC141355.1 | 8 | 1435 | ||
BC141356.1 | 8 | 1435 | ||
D30779.1 | 8 | 1507 | BAA06443.1 |