Assay Details
Target Gene Details
Entrez Gene ID: | 18779 |
Gene Name: | phospholipase A2 receptor 1 |
Gene Aliases: |
Pla2-r, Pla2g1br, Pla2r |
Location: |
Chr.2:60417543-60553350 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2r1 | NM_008867.2 | 17 | 2734 | NP_032893.1 |
BC141355.1 | 17 | 2615 | ||
BC141356.1 | 17 | 2615 | ||
D30779.1 | 17 | 2687 | BAA06443.1 |