Assay Details
Target Gene Details
Entrez Gene ID: | 99011 |
Gene Name: | protein-O-mannosyltransferase 1 |
Gene Aliases: |
AI505244 |
Location: |
Chr.2:32236595-32255005 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pomt1 | NM_145145.1 | 9 | 823 | NP_660127.1 |
AK053889.1 | 8 | 1343 | BAC35577.1 | |
AK134770.1 | 9 | 867 | BAE22274.1 | |
AK153984.1 | 9 | 922 | BAE32295.1 | |
AY007238.1 | 9 | 863 | AAG15588.1 | |
AY494857.1 | 9 | 883 | AAS76201.1 | |
BC027325.1 | 9 | 823 | AAH27325.1 |
More Information
Set Membership: |
Intragenic Exonic |