Assay Details
Target Gene Details
Entrez Gene ID: | 228026 |
Gene Name: | pyruvate dehydrogenase kinase, isoenzyme 1 |
Gene Aliases: |
B830012B01, D530020C15Rik |
Location: |
Chr.2:71873218-71903858 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pdk1 | NM_172665.5 | NP_766253.2 | ||
AK038454.1 | ||||
AK046805.1 | BAC32879.1 | |||
AK085215.1 | ||||
AK153649.1 | BAE32134.1 | |||
AK163553.1 | ||||
AK204344.1 | ||||
BC027196.1 | AAH27196.1 |
More Information
Set Membership: |
Intragenic Non-exonic |