Assay Details
Target Gene Details
Entrez Gene ID: | 18573 |
Gene Name: | phosphodiesterase 1A, calmodulin-dependent |
Gene Aliases: |
AI987702, AW125737 |
Location: |
Chr.2:79834453-80196518 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde1a | NM_001009978.1 | NP_001009978.1 | ||
NM_001009979.1 | NP_001009979.1 | |||
NM_001159582.1 | NP_001153054.1 | |||
NM_016744.4 | NP_058024.2 | |||
AF023529.1 | AAB81952.1 | |||
AF159298.1 | AAF63857.1 | |||
AK014810.1 | ||||
AK043647.1 | BAC31606.1 | |||
AK050004.1 | ||||
AY845863.1 | ||||
AY845864.1 | AAW31879.1 | |||
BC090628.1 | AAH90628.1 | |||
U56649.1 | AAB03319.1 |
More Information
Set Membership: |
Intragenic Non-exonic |