Assay Details
Target Gene Details
Entrez Gene ID: | 78928 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class T |
Gene Aliases: |
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7 |
Location: |
Chr.2:164497511-164508301 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigt | NM_133779.2 | NP_598540.2 | ||
AK019717.2 | ||||
AK044499.1 | BAC31951.1 | |||
AK049171.1 | ||||
AK049990.1 | ||||
AK153462.1 | BAE32014.1 | |||
AK153464.1 | BAE32015.1 | |||
AK155057.1 | BAE33018.1 | |||
AK157226.1 | BAE34008.1 | |||
AK177432.1 | ||||
BC034175.1 | AAH34175.1 |