Assay Details
Target Gene Details
Entrez Gene ID: | 18012 |
Gene Name: | neurogenic differentiation 1 |
Gene Aliases: |
BETA2, BHF-1, Neurod, bHLHa3 |
Location: |
Chr.2:79452641-79456636 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Neurod1 | NM_010894.2 | 1 | 43 | NP_035024.1 |
AK005073.1 | 1 | 44 | BAB23797.1 | |
AK018781.1 | 1 | 42 | BAB31405.1 | |
AK028055.1 | 1 | 47 | BAC25725.1 | |
BC018241.1 | AAH18241.1 | |||
BC094611.1 | AAH94611.1 |
More Information
Set Membership: |
Intragenic Exonic |