Assay Details
Target Gene Details
Entrez Gene ID: | 68349 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 3 |
Gene Aliases: |
0610010M09Rik, CI-30kD |
Location: |
Chr.2:90894636-90904721 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufs3 | NM_026688.2 | NP_080964.1 | ||
AK002501.1 | BAB22149.1 | |||
AK088337.1 | BAC40291.1 | |||
AK160888.1 | ||||
BC027270.1 | AAH27270.1 | |||
BC119267.1 | ||||
BC119269.1 |
More Information
Set Membership: |
Intragenic Non-exonic |