Assay Details
Target Gene Details
Entrez Gene ID: | 68375 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8 |
Gene Aliases: |
0610033L03Rik, AW261656 |
Location: |
Chr.2:36036334-36049292 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufa8 | NM_026703.2 | NP_080979.1 | ||
AK002744.1 | BAB22322.1 | |||
AK088463.1 | ||||
BC012416.1 | AAH12416.1 | |||
BC024673.1 | AAH24673.1 |
More Information
Set Membership: |
Intragenic Non-exonic |