Assay Details
Target Gene Details
Entrez Gene ID: | 56406 |
Gene Name: | nuclear receptor coactivator 6 |
Gene Aliases: |
AIB3, ASC-2, ASC2, NRC, Ncoa7, PRIP, RAP250, mKIAA0181 |
Location: |
Chr.2:155390656-155473894 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa6 | NM_001242558.1 | 4 | 592 | NP_001229487.1 |
NM_019825.3 | 4 | 592 | NP_062799.2 | |
AF216186.1 | 3 | 483 | AAF35860.1 | |
AK144338.1 | 6 | 831 | ||
AK171864.1 | 4 | 753 | BAE42705.1 | |
BC031113.1 | 4 | 596 | AAH31113.1 | |
BC083187.1 | 4 | 592 | AAH83187.1 | |
JF707634.1 | 3 | 451 | ||
JF707635.1 | 3 | 451 | ||
JF707636.1 | 3 | 451 |
More Information
Set Membership: |
Intragenic Exonic |