Assay Details
Target Gene Details
Entrez Gene ID: | 59126 |
Gene Name: | NIMA (never in mitosis gene a)-related expressed kinase 6 |
Gene Aliases: |
1300007C09Rik |
Location: |
Chr.2:38511697-38587490 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nek6 | NM_001159631.1 | NP_001153103.1 | ||
NM_021606.3 | NP_067619.1 | |||
AF218847.1 | AAG16653.1 | |||
AK004925.1 | BAB23676.2 | |||
AK011519.1 | BAB27673.2 | |||
AK018168.1 | ||||
AK029266.1 | ||||
AK075717.1 | BAC35907.1 | |||
AK075836.1 | BAC35995.1 | |||
AK089919.1 | BAC40995.1 | |||
AK170757.1 | BAE42008.1 | |||
BC019524.1 | AAH19524.1 |
More Information
Set Membership: |
Intragenic Non-exonic |