Assay Details
Target Gene Details
Entrez Gene ID: | 59030 |
Gene Name: | McKusick-Kaufman syndrome |
Gene Aliases: |
1300013E18Rik, AI463362, AI957237, Bbs6 |
Location: |
Chr.2:136873780-136891505 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkks | NM_001141946.1 | 2 | 248 | NP_001135418.1 |
NM_001286981.1 | 2 | 484 | NP_001273910.1 | |
NM_001286983.1 | 2 | 484 | NP_001273912.1 | |
NM_021527.2 | 2 | 484 | NP_067502.2 | |
AF254074.1 | 2 | 256 | AAF73965.1 | |
AK004987.1 | 2 | 241 | BAB23722.1 | |
AK008160.1 | 2 | 226 | ||
AK008406.1 | 2 | 226 | ||
AK032528.1 | 2 | 246 | BAC27912.1 | |
AK032554.1 | 2 | 246 | BAC27921.1 | |
BC024359.1 | 3 | 281 | AAH24359.1 | |
BY735964.1 | 2 | 249 |
More Information
Set Membership: |
Intragenic Exonic |