Assay Details
Target Gene Details
Entrez Gene ID: | 17005 |
Gene Name: | leukocyte tyrosine kinase |
Gene Aliases: |
- |
Location: |
Chr.2:119751326-119760431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltk | NM_008523.2 | NP_032549.2 | ||
NM_203345.2 | NP_976220.2 | |||
NM_206941.1 | NP_996824.1 | |||
NM_206942.2 | NP_996825.2 | |||
AK157478.1 | ||||
BC047138.1 | ||||
BC141299.1 | ||||
M90470.1 | AAA39451.1 | |||
X07984.1 | CAA30793.1 | |||
X52621.1 | CAA36848.1 |
More Information
Set Membership: |
Intragenic Non-exonic |