Assay Details
Target Gene Details
Entrez Gene ID: | 17153 |
Gene Name: | myelin and lymphocyte protein, T cell differentiation protein |
Gene Aliases: |
Mpv17, Vip17 |
Location: |
Chr.2:127633226-127656695 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mal | NM_001171187.1 | NP_001164658.1 | ||
NM_010762.5 | 2 | 699 | NP_034892.1 | |
AK004639.1 | 2 | 286 | BAB23430.1 | |
AK019046.1 | BAB31523.1 | |||
AK158653.1 | 2 | 700 | BAE34599.1 | |
AK169172.1 | 2 | 223 | BAE40951.1 | |
BC006826.1 | 2 | 196 | AAH06826.1 | |
Y07626.1 | 2 | 175 | CAA68907.1 | |
Y07812.1 | 2 | 137 | CAA69143.1 |
More Information
Set Membership: |
Intragenic Exonic |