Assay Details
Target Gene Details
Entrez Gene ID: | 16874 |
Gene Name: | LIM homeobox protein 6 |
Gene Aliases: |
Lhx6.1 |
Location: |
Chr.2:36081950-36105997 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lhx6 | NM_001083125.1 | 5 | 822 | NP_001076594.1 |
NM_001083126.1 | 4 | 632 | NP_001076595.1 | |
NM_001083127.1 | 4 | 632 | NP_001076596.1 | |
NM_008500.2 | 5 | 822 | NP_032526.2 | |
AB031039.1 | 4 | 632 | BAA83420.1 | |
AB031040.1 | 4 | 632 | BAA83421.1 | |
AJ000337.1 | 5 | 664 | CAA04011.1 | |
AK140987.1 | 5 | 823 | ||
BC065077.1 | 4 | 566 | AAH65077.1 |
More Information
Set Membership: |
Intragenic Exonic |