Assay Details
Target Gene Details
Entrez Gene ID: | 94217 |
Gene Name: | low density lipoprotein-related protein 1B (deleted in tumors) |
Gene Aliases: |
9630004P12Rik, LRP-DIT |
Location: |
Chr.2:40595290-42653631 on Build GRCm38 |
Assay Gene Location: | Within Exon 73 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lrp1b | NM_053011.2 | 73 | 11970 | NP_443737.2 |
AF270884.1 | 73 | 11970 | AAF81758.1 |
More Information
Set Membership: |
Intragenic Exonic |