Assay Details
Target Gene Details
Entrez Gene ID: | 16434 |
Gene Name: | inosine triphosphatase (nucleoside triphosphate pyrophosphatase) |
Gene Aliases: |
2010016I08Rik, AU020102, Itp |
Location: |
Chr.2:130667841-130681614 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itpa | NM_025922.2 | NP_080198.2 | ||
AB100501.1 | BAD04064.1 | |||
AK008279.1 | BAB25571.1 | |||
AK009248.1 | ||||
AK138101.1 | ||||
AK139770.1 | BAE24133.1 | |||
AK153806.1 | BAE32191.1 | |||
AK153904.1 | BAE32246.1 | |||
BC026508.1 | AAH26508.1 | |||
BC094466.1 | AAH94466.1 |
More Information
Set Membership: |
Intragenic Non-exonic |