Assay Details
Target Gene Details
Entrez Gene ID: | 16410 |
Gene Name: | integrin alpha V |
Gene Aliases: |
1110004F14Rik, 2610028E01Rik, CD51, D430040G12Rik |
Location: |
Chr.2:83723819-83806917 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgav | NM_008402.3 | 3 | 695 | NP_032428.2 |
AK020674.1 | 3 | 623 | ||
AK080357.1 | 3 | 634 | ||
AK149984.1 | 3 | 597 | ||
U14135.1 | 3 | 577 | AAC52497.1 |
More Information
Set Membership: |
Intragenic Exonic |