Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifih1 | NM_001164477.1 | NP_001157949.1 | ||
NM_027835.3 | 3 | 987 | NP_082111.2 | |
AF374384.1 | 3 | 896 | AAM21359.1 | |
AK037057.1 | 3 | 885 | BAC29687.2 | |
AK040519.1 | 3 | 944 | BAC30614.1 | |
AK149802.1 | 3 | 894 | BAE29095.1 | |
AK153018.1 | 3 | 894 | BAE31652.1 | |
AK156450.1 | 3 | 895 | BAE33716.1 | |
AK168646.1 | 3 | 988 | BAE40503.1 | |
AY075132.1 | 3 | 662 | AAL77205.1 | |
BC080200.1 | AAH80200.1 | |||
EF113547.1 | 3 | 676 | ||
EF113548.1 | 3 | 676 | ||
EU084750.1 | 3 | 676 |