Assay Details
Target Gene Details
Entrez Gene ID: | 71586 |
Gene Name: | interferon induced with helicase C domain 1 |
Gene Aliases: |
9130009C22Rik, Helicard, Hlcd, MDA5, RLR-2 |
Location: |
Chr.2:62595612-62646255 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ifih1 | NM_001164477.1 | 11 | 2553 | NP_001157949.1 |
NM_027835.3 | 12 | 2700 | NP_082111.2 | |
AF374384.1 | 12 | 2609 | AAM21359.1 | |
AK018602.1 | BAB31303.2 | |||
AK153018.1 | 12 | 2607 | BAE31652.1 | |
AK153344.1 | 4 | 634 | BAE31920.1 | |
AY075132.1 | 12 | 2375 | AAL77205.1 | |
BC004031.1 | 5 | 842 | AAH04031.1 | |
BC025508.1 | 7 | 1144 | AAH25508.1 | |
BC080200.1 | 11 | 2458 | AAH80200.1 | |
EF113547.1 | 12 | 2389 | ||
EF113548.1 | 12 | 2389 | ||
EU084750.1 | 12 | 2389 |