Assay Details
Target Gene Details
Entrez Gene ID: | 16403 |
Gene Name: | integrin alpha 6 |
Gene Aliases: |
5033401O05Rik, AI115430, Cd49f, VLA-6 |
Location: |
Chr.2:71786885-71858427 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga6 | NM_001277970.1 | 6 | 1035 | NP_001264899.1 |
NM_008397.4 | 6 | 1035 | NP_032423.2 | |
AK034186.1 | 6 | 892 | BAC28623.1 | |
AK159653.1 | BAE35264.1 | |||
BC049132.1 | 7 | 960 | AAH49132.1 | |
BC058095.1 | 7 | 960 | AAH58095.1 | |
X69902.1 | 6 | 1016 | CAA49527.1 |
More Information
Set Membership: |
Intragenic Exonic |