Assay Details
Target Gene Details
Entrez Gene ID: | 67136 |
Gene Name: | kelch repeat and BTB (POZ) domain containing 4 |
Gene Aliases: |
2510026C23Rik |
Location: |
Chr.2:90904724-90910561 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kbtbd4 | NM_025991.3 | NP_080267.1 | ||
AK010998.1 | BAB27317.1 | |||
AK014373.1 | BAB29305.1 | |||
BC025103.1 | AAH25103.1 |